Learn More
Bioworld IEF (Isoelectrofocusing) Anode Buffer (50X), 500 mL
Supplier:  Bioworld 105300031
Encompass_Preferred
Product DescriptionIEF anode buffer has a 50X concentration and this anode buffer is used for isoelectric focusing. It has a pH value of 2.4. A concentration of 1X contains 7 mM phosphoric acid, dilute this buffer with deionized water to make 1X. IEF is a sensitive technique that is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensures consistent results.
Catalog No. 50-196-653