Learn More
Medchemexpress LLC IEM 1925 dihydrobromide | 258282-23-4 | 98.0% | 422.24 | C17H30Br2N2 | 5 MG

Supplier: Medchemexpress LLC HY1032325MG
IEM-1925 dihydrobromide is the dihydrobromide salt of IEM-1925, a use- and voltage-dependent open-channel antagonist of AMPA (glutamate) receptors. It is supplied as a high-purity solid for research use in pharmacology, electrophysiology, and seizure-model studies.
- Chemical formula: C17H30Br2N2.
- Cas number: 258282-23-4.
- Molecular weight: 422.24.
- Purity: 98.0%.
- Appearance: solid.
- Typical applications: AMPA receptor pharmacology and seizure-model research.
- Available pack sizes: 5 mg, 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.