Learn More
Waters Corp IEX-MS Start Up Kit, BioResolve SCX mAb Column, 3 μm
Supplier:  Waters Corp 176004500
Encompass_Preferred
This IEX-MS Start Up Kit is designed for reproducible, high-resolution, charge-based separations of monoclonal antibodies (mAbs) and other proteins in LC and LC-MS applications. It includes a BioResolve SCX mAb, 3 μm, corrosion–resistant column, a Humanized mAb Mass Check Standard, and pH Buffer Concentrates for enhanced performance.
  • Advanced column technology features non-porous, polymer-based particles with sulfonic acid ligands for high-resolution ion exchange separations.
  • Corrosion resistance extends column lifespan and ensures consistent performance.
  • Comprehensive kit includes a BioResolve SCX mAb column (2.1 x 50 mm, 3 μm), Humanized mAb Mass Check Standard, and pH concentrates.
Catalog No. 50-239-1125