Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ifebemtinib hydrochloride | 99.2% | 625.01 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY122844A

Encompass_Preferred

Ifebemtinib hydrochloride is a laboratory chemical intended for the manufacture of substances and research use. It is supplied as a white to off-white solid. This product is for research purposes only and should be handled by qualified scientists in appropriately equipped facilities. It has not been fully validated for medical applications.

  • Identified as a laboratory chemical for research use
  • For manufacture of substances
  • Supplied as a white to off-white solid
  • Molecular formula: C28H29ClF4N6O4
  • Storage at 4°C, sealed, away from moisture and light

Catalog No. 50-004-38684


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.