Learn More
Selleck Chemical LLC Ifenprodil E4881-5MG
SDP
Supplier:  Selleck Chemical LLC E48815MG
Encompass_Preferred
Ifenprodil(Dilvax RC 61-91) is a potent antagonist of N-methyl-D-aspartate (NMDA) receptor that selectively inhibits receptors containing the NR2B subunit It exhibits potential for its use in the treatment of various neurological diseases
Catalog No. 50-000-43345