Learn More
Abcam Ifenprodil hemitartrate (mM/ml), NMDA antagonist, 10MM_ML
Supplier:  Abcam AB14667510MM_ML
Encompass_Preferred
MW 400.49 g/mol, Purity >99%. Non-competitive NMDA receptor antagonist acting at the polyamine site. Displays GluN2B (formerly NR2B) subunit selectivity. IC₅₀ values are 0.15 μM at NR1/NR2B and >30 μM for NR1/NR2A, NR1/NR2C and NR1/NR2D receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4531