Learn More
ABclonal Technology IFITM3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2433720UL
Encompass_Preferred
Interferon-induced transmembrane (IFITM) proteins are a family of interferon induced antiviral proteins The family contains five members including IFITM1 IFITM2 and IFITM3 and belong to the CD225 superfamily The protein encoded by this gene restricts cellular entry by diverse viral pathogens such as influenza A virus Ebola virus and Sars-CoV-2
Catalog No. 50-000-10554