Learn More
Aat Bioquest iFluor 647 PSA Imaging Kit with Goat Anti-Mouse IgG (100 Tests)
SDP
Supplier:  Aat Bioquest 45290
Encompass_Preferred
Power Styramide Signal Amplification PSA system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide TSA reagents. This catalog is for 100 Tests.
Catalog No. 50-225-0212