Learn More
Medchemexpress LLC IFN-alpha 13/IFNA13 100ug
SDP
Supplier:  Medchemexpress LLC HYP73123100UG
Encompass_Preferred
IFN-alpha 13 (IFNA13) belongs to type I interferon family is produced by macrophages with antiviral activities[1] IFN-alpha 13/IFNA13 Protein Cynomolgus (HEK293 His) contains 166 a a (C25-E190) produced in HEK293 cells with a C-terminal His-tag
Catalog No. 50-001-81230