Learn More
Medchemexpress LLC Interferon alpha-7 (IFNA7) protein, human, recombinant (P. pastoris), C-terminal His tag | >95.0% | 100 UG

Supplier: Medchemexpress LLC HYP76405100UG
Recombinant human interferon alpha-7 (IFN-α7) expressed in Pichia pastoris with a C-terminal His tag. The protein is 166 amino acids (C24-D189), supplied in PBS (pH 7.4), and >95% pure by reducing SDS-PAGE. Intended for research use in interferon signaling, antiviral assays, and biochemical characterization.
- Recombinant human interferon alpha-7 produced in P. pastoris.
- C-terminal His tag for detection and purification.
- 166 amino acids (C24-D189).
- Purity >95.0% as determined by reducing SDS-PAGE.
- Supplied in 0.2 μm filtered PBS, pH 7.4.
- Store at -80 °C and avoid repeated freeze-thaw cycles.
- Available in a 100 μg size for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.