Learn More
TARGETMOL CHEMICALS INC IFN ALPHA-IFNAR-IN-1 HYD 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1163010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IFN alpha-IFNAR-IN-1 hydrochloride is an inhibitor of the interaction between IFN-(alpha) and IFNAR. IFN alpha-IFNAR-IN-1 hydrochloride inhibits MVA-induced IFN-(alpha) responses by BM-pDCs with IC50 of 2-8 uM. purity: 99%
Catalog No. 50-257-7449