Learn More
CPC Scientific H-Cys-Leu-Lys-Asp-Arg-His-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LYMK001A
Encompass
The peptide IRRP1 is derived from a consensus IFN-alpha sequence and corresponds to a putative receptor recognition domain of IFN-alpha. The binding capacity of cells for IFN-alphas was increased in the presence of IRRP1. Increased receptor occupancy resulted in increased phosphorylation-activation of the transcription factor ISGF3 and enhanced antiviral activity.ONE-LETTER SEQUENCE: CLKDRHDMOLECULAR FORMULA: C35H59N13O12S1MOLECULAR WEIGHT:886STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [153840-64-3], RESEARCH AREA: MiscellaneousREFERENCES: E.N.Fish, FEBS Lett., 365, 87 (1995);
Catalog No. 50-210-9689