Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-epsilon Human 50ug
SDP

Supplier:  Medchemexpress LLC HYP7158950UG

Encompass_Preferred

Recombinant human interferon-epsilon (IFN-ε) bearing an N-terminal His tag, expressed in Escherichia coli and purified for laboratory research. This product supports studies of type I interferon signaling and mucosal host defense mechanisms, particularly in the female reproductive tract.

  • Recombinant protein expressed in E. coli.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Approximate molecular weight 26.1 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Available in small research amounts, including a 50 μg vial.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term preservation.
  • Activity not specified on the product page.

Catalog No. 50-001-95425


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.