Learn More
Medchemexpress LLC IFN-gamma Human Bi 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7814020UG
Encompass_Preferred
IFN-gamma Protein is a dimeric soluble cytokine It exerts antibacterial antiviral and antitumor effects through JAK-STAT mTOR MAPK and PI3K/AKT signaling pathways IFN-gamma Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived IFN-gamma protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93948