Learn More
Medchemexpress LLC IFN-gamma Mouse HE 500ug
SDP
Supplier:  Medchemexpress LLC HYP70667500UG
Encompass_Preferred
Recombinant mouse interferon-gamma (IFN-γ) produced in HEK293 cells and supplied as a lyophilized, tag-free protein for research use. The protein is full-length (mature sequence), exhibits expected biological activity in cell proliferation assays, and is supplied with storage and formulation guidance to support in vitro immunology and signaling studies.
  • Mammalian expression ensures native glycosylation and activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Demonstrated biological activity with ED50 values in the low ng/mL range.
  • Lyophilized formulation for enhanced stability and convenient shipping.
  • Available in multiple package sizes, including a 500 μg option.
  • Tag-free, full-length mature protein sequence provided for functional studies.
Catalog No. 50-001-84472