Learn More
Medchemexpress LLC Ifn-lambda 3/IL-28B protein, mouse (HEK293, His) | >95.0% | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7485150UG
Encompass_Preferred
Recombinant mouse IFN-lambda 3 (IL-28B) expressed in HEK293 cells with a C-terminal 6xHis tag. The protein (sequence D20-V193) is supplied as 50 μg of purified material and is intended for in vitro signaling assays, biological activity studies, and reagent development. Store and handle according to the manufacturer's SDS recommendations for proteins.
Catalog No. 50-002-92812