Learn More
Medchemexpress LLC IFNAR1 Cynomolgus 50ug

Supplier: Medchemexpress LLC HYP7095250UG
Recombinant IFNAR1 protein derived from cynomolgus macaque and produced in HEK293 cells, supplied lyophilized with a C-terminal 6xHis tag. The protein is provided at >95% purity and is intended for research use in type I interferon receptor biology, receptor-ligand interaction assays, and signaling studies.
- Expressed in HEK293 cells to support proper mammalian folding and post-translational modifications.
- C-terminal 6xHis tag for straightforward detection and purification.
- Greater than 95% purity by reducing SDS-PAGE for consistent experimental results.
- Lyophilized formulation from PBS (pH 7.4) for convenient storage and handling.
- Sequence range corresponds to XP_005548864 (A25-K437), enabling studies of native receptor domains.
- Recommended storage at -20°C with aliquoting to maintain stability and activity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.