Learn More
Selleck Chemical LLC IFNb2-A2975-1MG-25
SDP
Supplier:  Selleck Chemical LLC A29751MG25
Encompass_Preferred
IFNb2 is a humanized mouse monoclonal antibody that specifically binds to IL-6 and strongly inhibits IL-6 functions MW 145 46 KD
Catalog No. 50-313-5596
Add to Cart
Request a Quote