Learn More
Medchemexpress LLC Ifosfamide-d4 | 1189701-13-0 | 99.9% | 265.11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17419S11MG
Encompass_Preferred
Ifosfamide-d4 is a deuterium-labeled alkylating chemotherapeutic agent with activity against a wide range of tumors. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84306