Learn More
INTACT GENOMICS INC IG Chemically Competent Cells, Autoinduction DE3, E. coli, 96 x 50 μL
Supplier:  INTACT GENOMICS INC 106595
Encompass
Autoinduction DE3 Chemically Competent Cells are ideal for transformation and routine recombinant protein expression under an IPTG-inducible T7 promoter. This strain's chromosomally integrated IPTG-inducible promoter enables T7 RNA polymerase expression via IPTG induction or optimized autoinduction media. This system simplifies autoinduction, potentially increasing expressed cells and protein yields by 5 to 10 fold or more across diverse protein targets at various production scales.
  • Chemically competent cell type.
  • Autoinduction DE3 strain with chromosomally integrated IPTG-inducible promoter.
  • E. coli species.
  • Transformation efficiency of ≥ 3.0 x 107 cfu/μg pUC19 DNA.
  • Shipped on dry ice.
  • T7 expression strain.
  • Optimized for autoinduction.
  • Suitable for expression of toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
Catalog No. NC3998326