Learn More
Cayman Chemical IgA1 nd IgA2humn RabbIt 50ug
SDP
Supplier:  Cayman Chemical 3236850 UG
Encompass_Preferred
Immunogen: Human IgA • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgA1, IgA2; (-) Human IgG, IgM, IgD, and IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-295-0424