Learn More
Cayman Chemical IgA1humn RabbIt Monclona 50ug
SDP
Supplier:  Cayman Chemical 3236950 UG
Encompass_Preferred
Immunogen: Human IgA • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgA1, IgA2 (weakly); (-) Human IgG, IgM, IgD, IgE • Applications: ELISA, IHC, ICC
Catalog No. 50-294-6371