Learn More
Cayman Chemical IgA2humn Monclonal ntIb 100ug
SDP
Supplier:  Cayman Chemical 32116100 UG
Encompass_Preferred
Immunogen: Human IgA2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgA2 (-) Human IgA1, IgG, IgM, IgD, IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-295-0131