Learn More
Cayman Chemical IgAmouse Monclonal ntIbo 100ug
SDP
Supplier:  Cayman Chemical 20715100 UG
Encompass_Preferred
Immunogen: Mouse IgA • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Human IgA; (-) Mouse IgG1, IgG2a, IgG2b, IgG2c, IgG3, IgE, IgM • Applications: ELISA, WB (non-reducing conditions) • MW: ~160 kDa for non-reduced IgA • A probe for immunochemical detection of mouse IgA
Catalog No. 50-298-3192