Learn More
Enzo Life Sciences IgE (human) monoclonal antibody (I27) (biotin conjugate) (100µg)
Supplier: Enzo Life Sciences ENZABS2250100
Clone: I27. Isotype: IgG1. Immunogen: Purified human IgE (preincubated with mouse serum anti human IgG). Species reactivity: Human. Specificity: Recognizes human IgE (heat labile epitope in N-terminal of CH3 domain). Applications: Flow Cytometry, WB. Formulation: Liquid. In PBS containing 50% glycerol.