Learn More
Medchemexpress LLC HY-P7020 10ug Medchemexpress, IGF-BP3, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP702010UG
Encompass_Preferred
Medchemexpress, HY-P7020 10ug IGF-BP3, Human CAS: Recombinant Human Insulin-like Growth Factor-Binding Protein 3 belongs to insulin-like growth factor-binding protein, prolongs the half-life of the insulin-like growth factors (IGFs), inhibits interaction of IGFs with their cell surface receptors, and functions in cancer reduction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0553