Learn More
Medchemexpress LLC IGF-I/IGF-1 Human 5ug
SDP
Supplier:  Medchemexpress LLC HYP731365UG
Encompass_Preferred
The LR3 IGF-I/IGF-1 protein is similar to insulin and has potent growth-promoting activity that exceeds the efficacy of insulin As a potential physiological regulator it stimulates glucose transport and glycogen synthesis in osteoblasts even at significantly lower concentrations than insulin IGF-I/IGF-1 Protein Human (HEK293 hFc) is the recombinant human-derived IGF-I/IGF-1 protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80480