Learn More
Medchemexpress LLC IGF-I R Human Biot 100ug | 100ug
Supplier: Medchemexpress LLC HYP78142100UG
IGF-I receptor (IGF1R) mediates insulin-like growth factor effects binding strongly to IGF1 Activation triggers PI3K-AKT/PKB and Ras-MAPK pathways influencing cell survival protein synthesis and proliferation IGF1R also signals through JAK/STAT inhibiting JNK activation Hybrid receptors show variable binding to IGF1 and insulin IGF-I R Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived IGF-I R protein expressed by HEK293 with C-Avi C-His labeled tag