Learn More
BIOHIPPO IGF1A-V5/GFP
SDP
Supplier:  BIOHIPPO BHV194001111
Encompass
IGF1A-V5/GFP lentiviral vector expressing V5 under CMV promoter Each vial provides a nominal titer of 2x10E6 transducing units TU Human insulin like growth factor 1 (IGF1A) ORF cDNA lentivirus Lentiviral particles packaged with expression vector for human IGF1A (also known as IGF-1 somatomedin-C) transcript variant 4 A CMV promoter drives expression of IGF1A fused to a C-terminal V5 tag and an RFP reporter which is separated from the ORF cDNA by a self-cleaving peptide These lentiviral particles allow for easy transduction and high stable expression of the indicated ORF cDNA in mammalian cells The lentiviral vectors have been optimized to provide high gene expression and reliable genome integration allowing for easy establishment of long-term stable cell lines with consistent read out of fluorescent (GFP RFP) or luminescent (firefly luc) reporters
Catalog No. 50-003-31890