Learn More
Cayman Chemical IGF1RCD221 EXrAClular Do 100ug
SDP
Supplier:  Cayman Chemical 33982100 UG
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged IGF-1R expressed in HEK293 cells • Amino acids: 31-932 • MW: 81 (α subunit), 23 (β subunit), and 104 kDa (single chain)
Catalog No. 50-294-7270