Learn More
ABclonal Technology IGF2BP1/IMP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A25715100UL
Encompass_Preferred
This gene encodes a member of the insulin-like growth factor 2 mRNA-binding protein family The protein encoded by this gene contains four K homology domains and two RNA recognition motifs It functions by binding to the mRNAs of certain genes including insulin-like growth factor 2 beta-actin and beta-transducin repeat-containing protein and regulating their translation Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12538