Learn More
Medchemexpress LLC Recombinant human IGF2R protein, biotinylated | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70086950UG
Encompass_Preferred
Biotinylated recombinant human IGF2R protein (E47-M1508) is supplied as a lyophilized HEK293-expressed construct with C-terminal His and Avi tags for affinity purification and site-specific biotinylation. The product is intended for research on receptor-ligand interactions, intracellular trafficking, and related biochemical assays.
  • Biotinylated recombinant protein suitable for ligand binding and trafficking studies.
  • C-terminal His and Avi tags enable affinity purification and site-specific biotinylation.
  • Expressed in mammalian HEK293 cells to preserve native glycosylation.
  • Construct spans E47-M1508, covering relevant extracellular domains.
  • Glycosylated protein with an apparent molecular weight of approximately 165-200 kDa.
  • Lyophilized formulation with protectant; endotoxin level <1 EU/μg and recommended storage at -20°C.
Catalog No. 50-001-77841