Learn More
Medchemexpress LLC IGFBP-4 protein, mouse (HEK293, C-His) | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7484550UG
IGFBP-4 protein, mouse (HEK293, His) is a recombinant insulin-like growth factor-binding protein produced in mammalian cells and supplied as a 0.22 μm filtered PBS solution for research applications. The product is ~38 kDa, carries a C-terminal His tag, and is characterized for purity and activity to support biochemical and cell-based assays.
- Recombinant mouse IGFBP-4 expressed in HEK293 cells.
- C-terminal His tag facilitates purification and detection.
- Supplied in PBS, 0.22 μm filtered, for immediate use in assays.
- High purity (>95%) by reducing SDS-PAGE for reliable results.
- Molecular weight approximately 38 kDa.
- Biological activity with ED50 ~0.1-0.6 μg/mL in cell proliferation assays.
- Stable when stored at -80°C; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.