Learn More
ABclonal Technology IGFBP4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A150681000UL
Encompass_Preferred
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
Catalog No. 50-001-08295