Learn More
ABclonal Technology IGFBP5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A12451100UL
Encompass_Preferred
Enables insulin-like growth factor I binding activity. Involved in several processes, including cellular response to cAMP, regulation of smooth muscle cell migration, and regulation of smooth muscle cell proliferation. Part of insulin-like growth factor ternary complex. Biomarker of pulmonary fibrosis.
Catalog No. 50-001-30988