Learn More
ACROBiosystems Recombinant Protein;Human IGF-I R / CD221 Protein;HEK293;100UG;IGR,IGF1R,CD221,IGFIR,IGFR,JTK13,IGF-IR,IGF-1R,JTK-13,MGC142170,MGC142172,MGC18216,IGF-I R
Supplier:  ACROBiosystems IGRH5229100UG
Encompass_Preferred
Recombinant Protein;Human IGF-I R / CD221 Protein;100UG;Human IGF-I R (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession # P08069-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 104.8 kDa. The protein migrates as 90-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IGF-I R (Cat. No. IGR-H5229) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.04-0.313 μg/mL (QC tested).;Immobilized Human IGF-I R (Cat. No. IGR-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IGF-II, Fc Tag (Cat. No. IG2-H82F9) with a linear range of 0.156-5 μg/mL (Routinely tested).
Catalog No. 50-165-7557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.