Learn More
Cayman Chemical IgG Fabmouse Monclonal n 100ug
SDP
Supplier:  Cayman Chemical 21780100 UG
Encompass_Preferred
Immunogen: Mouse IgG • Host: Goat • Species Reactivity: (+) Mouse • Cross Reactivity: (+) Mouse IgG1, IgG2a, IgG2b, IgG3; (-) Mouse IgM, IgA, IgE; (-) Human, rabbit, rat IgG • Applications: ELISA
Catalog No. 50-298-7391