Learn More
Cayman Chemical IgG FcrabbIt Monclonal n 100ug
SDP
Supplier:  Cayman Chemical 32104100 UG
Encompass_Preferred
Immunogen: Rabbit IgG • Host: Goat • Species Reactivity: (+) Rabbit • Cross Reactivity: (-) Human, mouse, rat IgG • Applications: ELISA, WB (non-reducing conditions) • MW: ~150 kDa
Catalog No. 50-294-7533