Learn More
Athens Research And Technology Inc IGG HUMAN PLASMA 1GRAM
Supplier:  Athens Research And Technology Inc 16160907071GR
Encompass_Preferred
Immunoglobulin G, Human Plasma: 1 GRAM; Lyophilized in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 150,000; EC=1.36; Purity: > =95% by SDS-PAGE. Native; Purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-793