Learn More
Abcam Mouse IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1512761X96TESTS
Encompass_Preferred
Mouse IgG ELISA kit is a sandwich ELISA designed to quantify mouse immunoglobulin G (IgG) with a sensitivity of 2.2 ng/mL. - Colorimetric sandwich ELISA- 450nm readout - compatible with any standard plate reader - Wide dynamic range – 3.91–250 ng/mL - Broad sample compatibility – works with mouse serum, plasma, and cell culture supernatants - Cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0541131