Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG SepharoseR 6 fast flow
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17096901
Encompass
lgG Sepharose(R) 6 fast flow from Cytiva is based on the rigid Sepharose(R) 6 fast flow matrix with human IgG covalently coupled to it. Medium allows high flow rates to be used for rapid and convenient single step purification of protein.lgG Sepharose(R) 6 fast flow from Cytiva is based on the rigid Sepharose(R) 6 fast flow matrix with human IgG covalently coupled to it. The improved mechanical characteristics of this fast flow medium allows high flow rates to be used for rapid and convenient single step purification of protein A fusion protein conjungates produced in prokaryotic expression systems.The mechanical characteristics of this fast flow medium allows high flow rates to be used for rapid and convenient single step purification of protein A fusion protein conjungates produced in prokaryotic expression systems.
Catalog No. 50-290-4926