Learn More
Cayman Chemical IgG1humn RabbIt Monclona 50ug
SDP
Supplier:  Cayman Chemical 3237350 UG
Encompass_Preferred
Immunogen: Peptide from the hinge region of human IgG1 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG1; (-) Human IgG2, IgG3, IgG4, IgM, IgA, IgD, IgE (-) Goat, mouse, rat IgG • Applications: ELISA, ICC, IHC
Catalog No. 50-295-0192