Learn More
Cayman Chemical IgG2aKmouse RabbIt Monclo 50ug
SDP
Supplier:  Cayman Chemical 3235150 UG
Encompass_Preferred
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2aκ; (-) Mouse IgG2aλ, IgG1, IgG3, IgM, IgA, IgE; (-) Human, goat, rat IgG • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-8170