Learn More
Cayman Chemical IgG2amouse Monclonal ntI 100ug
SDP
Supplier:  Cayman Chemical 32005100 UG
Encompass_Preferred
Immunogen: Mouse IgG2a • Host: Rabbit • Species Reactivity: (+) Mouse IgG2a Fc region • Cross Reactivity: (–) Mouse IgG1, IgG2b, IgG2c, IgG3, IgM, IgA, IgE; (–) Human IgG; (–) Rat IgG • Applications: ELISA, FC, IP, WB (non-reducing conditions) • MW = ~150 kDa for non-reduced IgG2a
Catalog No. 50-294-8372