Learn More
Cayman Chemical IgG2bmouse RabbIt MoncLN 100ug
SDP
Supplier:  Cayman Chemical 32089100 UG
Encompass_Preferred
Immunogen: Mouse IgG2b • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Mouse IgG1, IgG2a, IgG3, IgM, IgA, IgE (-) Human, rat, goat IgG • Applications: ELISA, FC, IP, WB • MW: ~150 and 50 kDa for non-reduced and reduced IgG2b, respectively
Catalog No. 50-294-9162