Learn More
Medchemexpress LLC IgG3 Fc Human HEK 50ug
SDP
Supplier:  Medchemexpress LLC HYP7260050UG
Encompass_Preferred
The IgG3 Fc protein is the constant region of an immunoglobulin heavy chain that acts as a receptor for a specific antigen IgG3 Fc Protein Human (HEK293) is the recombinant human-derived IgG3 Fc protein expressed by HEK293 with tag free
Catalog No. 50-001-81036