Learn More
Cayman Chemical IgG3humn RabbIt Monclona 50ug
SDP
Supplier:  Cayman Chemical 3237550 UG
Encompass_Preferred
Immunogen: Human IgG3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG3; (-) Human IgG1, IgG2, IgG4, IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6584