Learn More
Cayman Chemical IgG3mouse RabbIt Monclona 50ug
SDP
Supplier:  Cayman Chemical 3235450 UG
Encompass_Preferred
Immunogen: Mouse IgG3 • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG3; (-) Mouse IgG1, IgG2a, IgG2b, IgM, IgA, IgE; (-) Human, rat IgG • Application: ELISA
Catalog No. 50-294-6919