Learn More
Medchemexpress LLC IgG4 Fc Human HEK2 10ug
SDP
Supplier:  Medchemexpress LLC HYP7077110UG
Encompass_Preferred
Recombinant human IgG4 Fc domain (E99-G326), expressed in HEK293 cells and supplied as a tag-free, lyophilized protein with mammalian glycosylation. Intended for antibody engineering, Fc functional assays, and assay development.
Catalog No. 50-001-96980