Learn More
Cayman Chemical IgG4humn RabbIt Monclona 50ug
SDP
Supplier:  Cayman Chemical 3214650 UG
Encompass_Preferred
Immunogen: Peptide corresponding to the hinge region of human IgG4 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG4; (-) Human IgG1, IgG2, IgG3, IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA, ICC, IHC, WB • MW = ~150 and 50 kDa for non-reduced and reduced IgG4, respectively
Catalog No. 50-295-0785